Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-iodo-7-deaza-2'-deoxyguanosine | 172163-62-1 | MFCD07778651; MFCD28134606 | 99.8% | 392.15 g·mol⁻¹ | C11H13IN4O4 | 250 MG
SDP

Catalog No. 5000209277
Encompass_Preferred

7-iodo-7-deaza-2'-deoxyguanosine is a halogenated deoxyguanosine nucleoside analog used as a building block in nucleic acid chemistry. It is used in oligonucleotide synthesis and sequencing, and can be converted to its triphosphate form for enzymatic incorporation.

  • High purity: 99.8%.
  • Chemical formula: C11H13IN4O4.
  • Molecular weight: 392.15 g·mol⁻¹.
  • Suitable for DNA synthesis and sequencing applications.
  • Can be triphosphorylated for enzymatic incorporation into DNA.
  • Available as a 250 mg package.

Catalog No. 50-002-09277 Supplier Medchemexpress LLC Supplier No. 250MGHYW048492
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-iodo-7-deaza-2'-deoxyguanosine is a halogenated deoxyguanosine nucleoside analog used as a building block in nucleic acid chemistry. It is used in oligonucleotide synthesis and sequencing, and can be converted to its triphosphate form for enzymatic incorporation.

  • High purity: 99.8%.
  • Chemical formula: C11H13IN4O4.
  • Molecular weight: 392.15 g·mol⁻¹.
  • Suitable for DNA synthesis and sequencing applications.
  • Can be triphosphorylated for enzymatic incorporation into DNA.
  • Available as a 250 mg package.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.