Learn More
Cayman Chemical 7-keto Cholesterol-d7, Purity: ≥99% deuterated forms (d1-d7), CAS: 127684-08-6, 5 mg
SDP
Catalog No. 502933484
Encompass_Preferred
An internal standard for the quantification of 7-keto cholesterol by GC- or LC-MS
Catalog No. 50-293-3484 Supplier Cayman Chemical Supplier No. 259725 MG
An internal standard for the quantification of 7-keto cholesterol by GC- or LC-MS