Learn More
Cayman Chemical 7-keto Deoxycholic Acid, Purity: ≥98%, CAS: 911-40-0, 50 mg
SDP
Catalog No. 502909522
Encompass_Preferred
A metabolite of cholic acid; fecal levels are elevated in rats fed a diet supplemented with cholic acid compared with rats fed a control diet or a diet supplemented with cholesterol; hepatic levels are elevated in a mouse model of ethanol-induced alcoholic liver disease
Catalog No. 50-290-9522 Supplier Cayman Chemical Supplier No. 3526450 MG
A metabolite of cholic acid; fecal levels are elevated in rats fed a diet supplemented with cholic acid compared with rats fed a control diet or a diet supplemented with cholesterol; hepatic levels are elevated in a mouse model of ethanol-induced alcoholic liver disease