Learn More
Medchemexpress LLC 7-keto-Deoxycholic acid | 911-40-0 | 99.7% | 406.56 | 1 ML
SDP
Catalog No. 5000404884
Encompass_Preferred
7-keto-Deoxycholic acid is a metabolite of bile acids, naturally found in *Clostridium absonum* and convertible by *Lactobacillus* and *Bifidobacterium*. It plays a role in the metabolism of cholic acid and its bioconversion rate increases with the presence of deoxycholic acid. This compound is for research use only.
  • Metabolite of bile acids
  • Convertible by *Lactobacillus* and *Bifidobacterium*
  • Exhibits increased bioconversion rate with deoxycholic acid
  • Suitable for research applications
  • Solid storage at 4°C, protected from light
  • Solution storage at -80°C for 6 months, or -20°C for 1 month, protected from light
Catalog No. 50-004-04884 Supplier Medchemexpress LLC Supplier No. HY4132410MM/1ML
7-keto-Deoxycholic acid is a metabolite of bile acids, naturally found in *Clostridium absonum* and convertible by *Lactobacillus* and *Bifidobacterium*. It plays a role in the metabolism of cholic acid and its bioconversion rate increases with the presence of deoxycholic acid. This compound is for research use only.
  • Metabolite of bile acids
  • Convertible by *Lactobacillus* and *Bifidobacterium*
  • Exhibits increased bioconversion rate with deoxycholic acid
  • Suitable for research applications
  • Solid storage at 4°C, protected from light
  • Solution storage at -80°C for 6 months, or -20°C for 1 month, protected from light