Learn More
Medchemexpress LLC 7-keto-Deoxycholic acid | 911-40-0 | 99.7% | 406.56 | 50 MG

Supplier: Medchemexpress LLC HY4132450MG
7-keto-Deoxycholic acid is a metabolite of bile acids found in *Clostridium absonum* and is also converted by *Lactobacillus* and *Bifidobacterium* under specific conditions. In vitro research shows that it exhibits an increased bioconversion rate with the addition of an equimolar concentration of deoxycholic acid, suggesting it is induced and subsequently metabolized. For example, *E. lentum*-like c-25 has been shown to convert cholic acid to deoxycholic acid and 7-keto-Deoxycholic acid during anaerobic incubation.
- Metabolite of bile acids in *Clostridium absonum*
- Converted by *Lactobacillus* and *Bifidobacterium*
- Bioconversion rate increases with deoxycholic acid
- Soluble in DMSO at 100 mg/mL
- Store at 4°C, protected from light
- In solvent, store at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.