Learn More
Medchemexpress LLC 7-keto-Deoxycholic acid | 911-40-0 | 99.71% | 406.56 | 25 MG
SDP
Catalog No. 5000403394
Encompass_Preferred
7-keto-Deoxycholic acid is a metabolite of bile acids found in *Clostridium absonum*. It can also be converted from *Lactobacillus* and *Bifidobacterium* under specific conditions. It exhibits an increased bioconversion rate when an equimolar concentration of deoxycholic acid is added.
  • Induced by deoxycholic acid and subsequently metabolized
  • *E. lentum-like c-25* converts cholic acid to deoxycholic acid and 7-keto-Deoxycholic acid under anaerobic incubation
Catalog No. 50-004-03394 Supplier Medchemexpress LLC Supplier No. 25MGHY41324R
7-keto-Deoxycholic acid is a metabolite of bile acids found in *Clostridium absonum*. It can also be converted from *Lactobacillus* and *Bifidobacterium* under specific conditions. It exhibits an increased bioconversion rate when an equimolar concentration of deoxycholic acid is added.
  • Induced by deoxycholic acid and subsequently metabolized
  • *E. lentum-like c-25* converts cholic acid to deoxycholic acid and 7-keto-Deoxycholic acid under anaerobic incubation