Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-keto-deoxycholic acid | 911-40-0 | MFCD00271373 | 99.5% | 406.56 | C24H38O5 | 10 MG
SDP

Catalog No. 5000299060
Encompass_Preferred

7-keto-Deoxycholic acid (Standard) is an analytical reference standard of the bile acid metabolite 7-keto-deoxycholic acid for research and analytical applications, including method development, HPLC, and mass spectrometry. It is supplied as a high-purity solid with documented solubility and defined storage conditions.

  • Analytical standard for metabolite identification and quantification.
  • High purity suitable for analytical workflows (assay and HPLC measurements reported).
  • Molecular formula C24H38O5 and molecular weight 406.56 provide exact mass reference.
  • Available in small solid packages and solution formats for flexible use.
  • Soluble in DMSO (requires sonication) and formulatable into in vivo vehicle preparations achieving ≥ 2.5 mg/mL.
  • Store solid protected from light at 4°C; store solutions frozen (-80°C for long term, -20°C short term).

Catalog No. 50-002-99060 Supplier Medchemexpress LLC Supplier No. HY41324R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-keto-Deoxycholic acid (Standard) is an analytical reference standard of the bile acid metabolite 7-keto-deoxycholic acid for research and analytical applications, including method development, HPLC, and mass spectrometry. It is supplied as a high-purity solid with documented solubility and defined storage conditions.

  • Analytical standard for metabolite identification and quantification.
  • High purity suitable for analytical workflows (assay and HPLC measurements reported).
  • Molecular formula C24H38O5 and molecular weight 406.56 provide exact mass reference.
  • Available in small solid packages and solution formats for flexible use.
  • Soluble in DMSO (requires sonication) and formulatable into in vivo vehicle preparations achieving ≥ 2.5 mg/mL.
  • Store solid protected from light at 4°C; store solutions frozen (-80°C for long term, -20°C short term).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.