Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-keto-Deoxycholic acid | 911-40-0 | MFCD00271373 | 99.7% | 406.56 | 100 MG
SDP

Catalog No. 5000403704
Encompass_Preferred

7-keto-Deoxycholic acid is a metabolite of bile acids found in *Clostridium absonum*. It can also be converted from *Lactobacillus* and *Bifidobacterium* under specific conditions.

  • High purity of 99.71%.
  • Molecular weight of 406.56.
  • Appears as a white to off-white solid.
  • Classified as ketones, aldehydes, and acids.
  • Endogenous metabolite of human origin.
  • Recommended storage at 4°C, protected from light.
  • Soluble in DMSO at 100 mg/mL with ultrasonication.

Catalog No. 50-004-03704 Supplier Medchemexpress LLC Supplier No. HY41324100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-keto-Deoxycholic acid is a metabolite of bile acids found in *Clostridium absonum*. It can also be converted from *Lactobacillus* and *Bifidobacterium* under specific conditions.

  • High purity of 99.71%.
  • Molecular weight of 406.56.
  • Appears as a white to off-white solid.
  • Classified as ketones, aldehydes, and acids.
  • Endogenous metabolite of human origin.
  • Recommended storage at 4°C, protected from light.
  • Soluble in DMSO at 100 mg/mL with ultrasonication.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.