Learn More
Cayman Chemical 7-keto Lithocholic Acid, Purity: ≥98%, CAS: 4651-67-6, 1 g
SDP
Catalog No. 502989077
Encompass_Preferred
An intermediate in the synthesis of ursodeoxycholic acid by intestinal bacteria in humans; increases the bile flow rate and the concentration of bile bicarbonate in rats via venous infusion at 1.2 µmol/min per 100 g body weight; levels are decreased in rat feces in a model of adenine-induced chronic renal failure; fecal levels are also reduced in patients with liver cirrhosis or hepatocellular carcinoma
Catalog No. 50-298-9077 Supplier Cayman Chemical Supplier No. 295461 G
An intermediate in the synthesis of ursodeoxycholic acid by intestinal bacteria in humans; increases the bile flow rate and the concentration of bile bicarbonate in rats via venous infusion at 1.2 µmol/min per 100 g body weight; levels are decreased in rat feces in a model of adenine-induced chronic renal failure; fecal levels are also reduced in patients with liver cirrhosis or hepatocellular carcinoma