Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-ketolithocholic acid (3α-hydroxy-7-oxo-5β-cholanic acid) | 4651-67-6 | MFCD00271393 | 99.9% | 390.56 g·mol⁻1 | C24H38O4 | 1 ML
SDP

Catalog No. 5000301022
Encompass_Preferred

7-Ketolithocholic acid (3α-hydroxy-7-oxo-5β-cholanic acid) is a bile acid derivative used to investigate bile acid metabolism, absorption, and regulation of biliary cholesterol secretion. It is provided as a high-purity solid or as a ready-to-use 10 mM solution in DMSO for in vitro and biochemical studies.

  • High purity: 99.93%.
  • Available as 10 mM solution in DMSO or as solid for custom formulation.
  • Molecular formula C24H38O4; molecular weight 390.56 g·mol⁻1.
  • Suitable for studies of bile acid signaling and metabolic pathways.
  • Intended for research use only.

Catalog No. 50-003-01022 Supplier Medchemexpress LLC Supplier No. HYW01851210MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Ketolithocholic acid (3α-hydroxy-7-oxo-5β-cholanic acid) is a bile acid derivative used to investigate bile acid metabolism, absorption, and regulation of biliary cholesterol secretion. It is provided as a high-purity solid or as a ready-to-use 10 mM solution in DMSO for in vitro and biochemical studies.

  • High purity: 99.93%.
  • Available as 10 mM solution in DMSO or as solid for custom formulation.
  • Molecular formula C24H38O4; molecular weight 390.56 g·mol⁻1.
  • Suitable for studies of bile acid signaling and metabolic pathways.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.