Learn More
Medchemexpress LLC 7-ketolithocholic acid | 4651-67-6 | MFCD00271393 | 99.9% | 390.56 g/mol | C24H38O4 | 5 G
SDP
Catalog No. 5000301285
Encompass_Preferred
7-Ketolithocholic acid is a bile acid research standard (3α-hydroxy-7-oxo-5β-cholanic acid) used in biochemical and metabolic studies. Supplied as a solid or as a DMSO solution, it is intended for in vitro research applications investigating bile acid metabolism and cholesterol secretion.
  • High purity suitable for research use.
  • White to off-white solid appearance.
  • Soluble in DMSO (≥ 100 mg/mL).
  • Stable under recommended storage conditions.
  • Includes molecular formula and molecular weight for analytical reference.
Catalog No. 50-003-01285 Supplier Medchemexpress LLC Supplier No. HYW0185125G
7-Ketolithocholic acid is a bile acid research standard (3α-hydroxy-7-oxo-5β-cholanic acid) used in biochemical and metabolic studies. Supplied as a solid or as a DMSO solution, it is intended for in vitro research applications investigating bile acid metabolism and cholesterol secretion.
  • High purity suitable for research use.
  • White to off-white solid appearance.
  • Soluble in DMSO (≥ 100 mg/mL).
  • Stable under recommended storage conditions.
  • Includes molecular formula and molecular weight for analytical reference.