Learn More
Medchemexpress LLC 7-ketolithocholic acid | 4651-67-6 | MFCD00271393 | 99.9% | 390.6 g/mol | C24H38O4 | 50 MG
SDP
Catalog No. 5000301296
Encompass_Preferred
7-ketolithocholic acid (3α-hydroxy-7-oxo-5β-cholanic acid) is a bile acid derivative supplied as an analytical standard for research and analytical applications. The material is provided as a high-purity solid suitable for use in chromatographic, metabolic, and quantitative studies.
  • High purity for reliable analytical results.
  • Suitable as a chromatographic reference standard.
  • Supports quantitative and metabolic profiling studies.
  • Supplied as a stable solid for convenient storage.
  • Available in small vial sizes for trace-level analyses.
  • CAS number 4651-67-6 provided for unambiguous identification.
Catalog No. 50-003-01296 Supplier Medchemexpress LLC Supplier No. HYW018512R50MG
7-ketolithocholic acid (3α-hydroxy-7-oxo-5β-cholanic acid) is a bile acid derivative supplied as an analytical standard for research and analytical applications. The material is provided as a high-purity solid suitable for use in chromatographic, metabolic, and quantitative studies.
  • High purity for reliable analytical results.
  • Suitable as a chromatographic reference standard.
  • Supports quantitative and metabolic profiling studies.
  • Supplied as a stable solid for convenient storage.
  • Available in small vial sizes for trace-level analyses.
  • CAS number 4651-67-6 provided for unambiguous identification.