Learn More
Cayman Chemical 7-Ketositosterol, Purity: ≥98%, CAS: 2034-74-4, 50 mg
SDP
Catalog No. 502919385
Encompass_Preferred
A phytosterol and phytosterol oxidation product; a thermal oxidation product of sitosterol; inhibits the proliferation of HL-60 cells (IC50 = 45.6 µM)
Catalog No. 50-291-9385 Supplier Cayman Chemical Supplier No. 3718950 MG
A phytosterol and phytosterol oxidation product; a thermal oxidation product of sitosterol; inhibits the proliferation of HL-60 cells (IC50 = 45.6 µM)