Learn More
K2 Scientific 7 kWh Battery Backup
SDP
Catalog No. VLUPSUS7KW
Change view
Click to view available options
Accessory
Catalog No.
VLUPSUS7KW Accessory
1 options
Catalog No. VLUPSUS7KW Supplier K2 Scientific Supplier No. VLUPSUS7KW
Add to Cart
Add to Cart
Request a Quote
7 Battery Backup with safer, longer lasting LiFePO4 batteries
The solution to safeguard your commercial equipment from power outages. With zero millisecond transfer time, this UPS ensures uninterrupted power, providing you with the peace of mind you deserve.
Key Features:
  • Zero Millisecond Transfer Time:Experience seamless power transitions, ensuring your critical equipment stays online without interruption.
  • Vital Power Log Technology:Stay informed and in control with our cutting-edge Vital Power Log technology. Receive real-time alerts and access detailed data on your battery status, ensuring you're always aware of your power situation.
  • Compatibility with Life ScienceCold Storage:Our 7KW Battery Backup seamlessly integrates with K2 units, as illustrated in the accompanying chart. This synergy enhances overall performance and reliability.
  • Connectivity:Receive text and email notifications, delivering real-time updates on run times, charge levels, and energy consumption usage. Stay informed and in control with timely alerts for optimal power management.
  • Reliable Performance:Trust the consistent and reliable power supply to protect your valuable equipment.
  • User-Friendly Interface:Easily monitor and manage your power situation with our intuitive interface, providing you with the control you need.
Specifications
Type Battery Backup
Exterior Finish Painted Aluminum