Learn More
Chemglass Life Sciences Agitator Assembly for 7L Bio Reactors, 8mm OD Shaft, M30 Thread
SDP
Supplier:  Chemglass Life Sciences CLS138304
Encompass
Agitator assembly is constructed of 316L stainless steel and screws into the M30 thread in the center of the headplate.
Catalog No. NC1937658