Learn More
Vetequip Inc 7 LITER INDUCTIN CHAMBER
SDP
Catalog No. NC9889337
Encompass_Preferred
Induction Chamber, 7 L, 8.25 W x 9 D x 5.75 in.H, Positive Sealing Gasket at Lid, Duarable, Clear Acrylic Construction, Non-skid Feet, Designed for Extra Large Rats, Ferrets and Small to Medium Rabbits
Catalog No. NC9889337 Supplier Vetequip Inc Supplier No. 941448
Induction Chamber, 7 L, 8.25 W x 9 D x 5.75 in.H, Positive Sealing Gasket at Lid, Duarable, Clear Acrylic Construction, Non-skid Feet, Designed for Extra Large Rats, Ferrets and Small to Medium Rabbits