Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-MAD-MDCPT hydrochloride | 2740593-09-1 | 457.86 g/mol | C22H20ClN3O6 | 5 MG
SDP

Catalog No. 5000262597
Encompass_Preferred

7-MAD-MDCPT hydrochloride is a camptothecin analog used as a cytotoxic payload in antibody-drug conjugates for research applications. Supplied as the hydrochloride salt in small laboratory quantities, it requires low-temperature, moisture-free storage. Soluble in DMSO; consult safety and handling documentation before use.

  • Camptothecin analog suitable as an ADC toxin payload.
  • Hydrochloride salt form for improved handling.
  • Molecular formula C22H20ClN3O6; molecular weight 457.86 g/mol.
  • CAS number 2740593-09-1 for unambiguous identification.
  • DMSO solubility approximately 6.67 mg/mL (14.57 mM).
  • Store at -20°C under inert gas; in solution store at -80°C for long-term.

Catalog No. 50-002-62597 Supplier Medchemexpress LLC Supplier No. HY132162A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-MAD-MDCPT hydrochloride is a camptothecin analog used as a cytotoxic payload in antibody-drug conjugates for research applications. Supplied as the hydrochloride salt in small laboratory quantities, it requires low-temperature, moisture-free storage. Soluble in DMSO; consult safety and handling documentation before use.

  • Camptothecin analog suitable as an ADC toxin payload.
  • Hydrochloride salt form for improved handling.
  • Molecular formula C22H20ClN3O6; molecular weight 457.86 g/mol.
  • CAS number 2740593-09-1 for unambiguous identification.
  • DMSO solubility approximately 6.67 mg/mL (14.57 mM).
  • Store at -20°C under inert gas; in solution store at -80°C for long-term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.