Learn More
AdipoGen 7-MeO-coumarin-carboxylic acid
SDP
Supplier:  AdipoGen CDXM0016G005
Encompass_Preferred
Chemical. CAS 20300-59-8. Formula C11H8O5. MW 220.18. Synthetic. An amine-reactive blue fluorescent probe that provides a distinguishable contrast when used with longer-wavelength probes in certain multicolor fluorescence applications such as nucleic acid and protein microarrays, in situ hybridization and immunofluorescence. This carboxylic acid derivative of 7-methoxycoumarin can be used for coupling to amines, hydrazines or hydroxylamines on biomolecules of interest. Spectral data Ex=336nm, Em=402nm Buffer, pH9.
Catalog No. 50-256-0572