Learn More
AdipoGen 7-MeO-coumarin-carboxylic NSE
SDP
Supplier:  AdipoGen CDXM0017M125
Encompass_Preferred
Chemical. CAS 150321-92-9. Formula C15H11NO7. MW 317.25. Synthetic. Excellent amino-reactive tag that has strong blue fluorescence. Can be used to create blue-fluorescent bioconjugates with an excitation/emission spectra of ~358/410 nm. Used to label neuropeptides, peptides, nucleotides and cells.
Catalog No. 50-256-0569