Learn More
Medchemexpress LLC 7-Methoxy-1-tetralone | 6836-19-7 | 176.21 | 25 G
SDP
Catalog No. 5000450023
Encompass_Preferred
7-Methoxy-1-tetralone is a potent antitumor agent that inhibits cancer cell proliferation and migration. It induces apoptosis in hepatocellular carcinoma (HCC) cells and has been shown to decrease the protein levels of NF-κB, matrix metallopeptidase 2 (MMP2)/MMP9, and p-AKT.
  • Inhibits cancer cell proliferation and migration
  • Induces apoptosis in hepatocellular carcinoma (HCC) cells
  • Decreases protein levels of NF-κB, matrix metallopeptidase 2 (MMP2)/MMP9, and p-AKT
  • Demonstrates antitumor activity in nude mice without affecting body weight or organ index
Catalog No. 50-004-50023 Supplier Medchemexpress LLC Supplier No. 25GHYW001925
7-Methoxy-1-tetralone is a potent antitumor agent that inhibits cancer cell proliferation and migration. It induces apoptosis in hepatocellular carcinoma (HCC) cells and has been shown to decrease the protein levels of NF-κB, matrix metallopeptidase 2 (MMP2)/MMP9, and p-AKT.
  • Inhibits cancer cell proliferation and migration
  • Induces apoptosis in hepatocellular carcinoma (HCC) cells
  • Decreases protein levels of NF-κB, matrix metallopeptidase 2 (MMP2)/MMP9, and p-AKT
  • Demonstrates antitumor activity in nude mice without affecting body weight or organ index