Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Methoxy-1-tetralone, 99%, Thermo Scientific™

Catalog No. AAL1513003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1513003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1513003 Supplier Thermo Scientific Chemicals Supplier No. L1513003

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6836-19-7
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD00001696
InChI Key GABLTKRIYDNDIN-UHFFFAOYSA-N
Synonym 7-methoxy-1-tetralone, 7-methoxyl-1-tetralone, 7-methoxy-1,2,3,4-tetrahydronaphthalen-1-one, 7-methoxy-3,4-dihydronaphthalen-1 2h-one, 1 2h-naphthalenone, 3,4-dihydro-7-methoxy, 7-methoxytetralin-1-one, 3,4-dihydro-7-methoxy-1 2h-naphthalenone, 7-methoxy-2,3,4-trihydronaphthalen-1-one, 7-methoxytetralone, 7-methoxy-tetralone
PubChem CID 81276
IUPAC Name 7-methoxy-3,4-dihydro-2H-naphthalen-1-one
SMILES COC1=CC2=C(CCCC2=O)C=C1

Specifications

Melting Point 59°C to 63°C
Quantity 1 g
Beilstein 2208389
Formula Weight 176.22
Percent Purity 99%
Chemical Name or Material 7-Methoxy-1-tetralone
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 229-916-0
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.