Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Methoxy-2-tetralone, TCI America™
SDP

Catalog No. M142125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M142125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M142125G Supplier TCI America Supplier No. M142125G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4133-34-0
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD00001730
InChI Key XEAPZXNZOJGVCZ-UHFFFAOYSA-N
Synonym 7-methoxy-2-tetralone, 7-methoxy-3,4-dihydronaphthalen-2 1h-one, 3,4-dihydro-7-methoxy-2 1h-naphthalenone, 7-methoxy-3,4-dihydronaphthalen-2-one, 7-methoxy-1,2,3,4-tetrahydronaphthalen-2-one, 7-methoxyl-2-tetralone, 7-methoxytetralin-2-one, 2 1h-naphthalenone, 3,4-dihydro-7-methoxy, 7-methoxy-1,3,4-trihydronaphthalen-2-one, 3,4-dihydro-7-methoxynaphthalen-2 1h-one
PubChem CID 77785
IUPAC Name 7-methoxy-3,4-dihydro-1H-naphthalen-2-one
SMILES COC1=CC2=C(CCC(=O)C2)C=C1

Specifications

Color Yellow
Quantity 25 g
Formula Weight 176.22
Percent Purity ≥95.0% (GC)
Physical Form Crystals
Chemical Name or Material 7-Methoxy-2-tetralone
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.