Learn More
AdipoGen 7-Methoxy-4-CF3 coumarin
SDP
Supplier:  AdipoGen CDXM0015M250
Encompass_Preferred
Chemical. CAS 575-04-2. Formula C11H7F3O3. MW 244.17. Synthetic. Fluorescent substrate for oxidoreductases, especially for Cytochrome P450.
Catalog No. 50-256-0573