Learn More
AdipoGen 7-Methoxy-4-methylcoumarin (100 mg)
SDP
Supplier:  AdipoGen CDXM0046M100
Encompass_Preferred
7-Methoxy-4-methylcoumarin (100 mg), AdipoGen Life Sciences. Chemical. CAS: 2555-28-4. Formula: C11H10O3. MW: 190.2. Synthetic. Coumarin derivative. Fluorescent label. Shown to be UVA-phototoxic against F. cultnorum and to have some weak biological activities.
Catalog No. 50-149-0485