Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-Methoxy-4-methyl-coumarin-8-ol | 22084-94-2 | 206.19 | 5 MG
SDP

Catalog No. 5000416794
Encompass_Preferred

7-Methoxy-4-methyl-coumarin-8-ol is a compound isolated from sour orange (Citrus aurantium). It is for research use only and not sold to patients. This product appears as a white to off-white solid and boasts a purity of 99.92%.

  • Isolated from sour orange (Citrus aurantium)
  • High purity of 99.92%
  • Appears as a white to off-white solid
  • For research use only
  • Ships at room temperature in continental US
  • Store at 4°C, sealed, away from moisture and light
  • Soluble in DMSO for in vitro applications
  • Soluble in 10% DMSO, 40% PEG300, 5% Tween-80, 45% Saline for in vivo applications

Catalog No. 50-004-16794 Supplier Medchemexpress LLC Supplier No. HYN100155MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Methoxy-4-methyl-coumarin-8-ol is a compound isolated from sour orange (Citrus aurantium). It is for research use only and not sold to patients. This product appears as a white to off-white solid and boasts a purity of 99.92%.

  • Isolated from sour orange (Citrus aurantium)
  • High purity of 99.92%
  • Appears as a white to off-white solid
  • For research use only
  • Ships at room temperature in continental US
  • Store at 4°C, sealed, away from moisture and light
  • Soluble in DMSO for in vitro applications
  • Soluble in 10% DMSO, 40% PEG300, 5% Tween-80, 45% Saline for in vivo applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.