Learn More
Selleck Chemical LLC 7-Methoxy-4-methylcoumarin S4780-25mg
SDP
Catalog No. 503111967
Encompass_Preferred
7-Methoxy-4-methylcoumarin is a coumarin derivative and fluorescent label 7-Methoxy-4-methylcoumarin has an antimicrobial activitiy against both gram positive and gram negative bacterial stains 4-Methylherniarin displays good activity against B subtilis and S sonnei with IC50 of 11 76 g/ml and 13 47 g/ml respectively
Catalog No. 50-311-1967 Supplier Selleck Chemical LLC Supplier No. S478025MG
7-Methoxy-4-methylcoumarin is a coumarin derivative and fluorescent label 7-Methoxy-4-methylcoumarin has an antimicrobial activitiy against both gram positive and gram negative bacterial stains 4-Methylherniarin displays good activity against B subtilis and S sonnei with IC50 of 11 76 g/ml and 13 47 g/ml respectively