Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7-Methoxy-4-methylcoumarin 98.0+%, TCI America™
SDP

Catalog No. M139325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M139325G 25 g
M13935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M139325G Supplier TCI America Supplier No. M139325G

Chemical Identifiers

CAS 2555-28-4
Molecular Formula C11H10O3
Molecular Weight (g/mol) 190.198
MDL Number MFCD00009773
InChI Key UDFPKNSWSYBIHO-UHFFFAOYSA-N
Synonym 7-methoxy-4-methylcoumarin, 7-methoxy-4-methyl-2h-chromen-2-one, 2h-1-benzopyran-2-one, 7-methoxy-4-methyl, 4-methylherniarin, hymecromone methyl ether, herniarin, 4-methyl, 7-methoxy-4-methyl-chromen-2-one, 4-methyl-7-methoxycoumarin, coumarin,4-methyl-7-methoxy, coumarin, 7-methoxy-4-methyl
PubChem CID 390807
IUPAC Name 7-methoxy-4-methylchromen-2-one
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)OC

Specifications

Melting Point 160°C
Color White-Yellow
Quantity 25 g
Formula Weight 190.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 7-Methoxy-4-methylcoumarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.