Learn More
Abcam 7-Metho x y-4-(trifluoromethyl)coumarin, Fluorescent substrate for o x idoreductases, 250MG
Catalog No. 502705201
Encompass_Preferred
MW 244.17 g/mol, Purity >99%. Fluorescent substrate for oxidoreductases, especially for Cytochrome P450.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5201 Supplier Abcam Supplier No. AB145426250MG
MW 244.17 g/mol, Purity >99%. Fluorescent substrate for oxidoreductases, especially for Cytochrome P450.
The product is subject to the following: Abcam Restricted Use Statement