Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-methoxybenzo[d][1,3]dioxol-5-ol | 50901-34-3 | 96.0% | 168.15 | C8H8O4 | 10 MG
SDP

Catalog No. 5000210406
Encompass_Preferred

Antioxidant agent-4 is a small-molecule antioxidant reported to exhibit antilipid peroxidative activity. It is supplied for research use and is characterized by its chemical identifiers, purity, and storage stability.

  • Chemical name 7-methoxybenzo[d][1,3]dioxol-5-ol.
  • Cas number 50901-34-3.
  • Molecular formula C8H8O4.
  • Molecular weight 168.15.
  • Purity 96.0%.
  • Storage and stability in solvent: -80°C for 6 months, -20°C for 1 month when stored under nitrogen.
  • Typical package sizes include 10 mg and solution preparations.

Catalog No. 50-002-10406 Supplier Medchemexpress LLC Supplier No. HYW06842010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antioxidant agent-4 is a small-molecule antioxidant reported to exhibit antilipid peroxidative activity. It is supplied for research use and is characterized by its chemical identifiers, purity, and storage stability.

  • Chemical name 7-methoxybenzo[d][1,3]dioxol-5-ol.
  • Cas number 50901-34-3.
  • Molecular formula C8H8O4.
  • Molecular weight 168.15.
  • Purity 96.0%.
  • Storage and stability in solvent: -80°C for 6 months, -20°C for 1 month when stored under nitrogen.
  • Typical package sizes include 10 mg and solution preparations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.