Learn More
Selleck Chemical LLC 7-Methoxycoumarin S5163-25mg
SDP
Catalog No. 503112109
Encompass_Preferred
7-Methoxycoumarin (Herniarine) also known as Herniarin is a methoxy derivative of coumarin and substrate for dealkylase It has been used in the synthesis of anti-first-pass effect compounds and detergents
Catalog No. 50-311-2109 Supplier Selleck Chemical LLC Supplier No. S516325MG
7-Methoxycoumarin (Herniarine) also known as Herniarin is a methoxy derivative of coumarin and substrate for dealkylase It has been used in the synthesis of anti-first-pass effect compounds and detergents