Learn More
Abcam 7-Metho x ycoumarin-3-carbo x ylic acid, Amine-reactive fluorescent probe, 500MG
Catalog No. 502705186
Encompass_Preferred
MW 220.18 g/mol, Purity >98%. Amine-reactive fluorescent probe. Used for coupling to amines, hydrazines or hydroxylamines on biomolecules of interest.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5186 Supplier Abcam Supplier No. AB145444500MG
MW 220.18 g/mol, Purity >98%. Amine-reactive fluorescent probe. Used for coupling to amines, hydrazines or hydroxylamines on biomolecules of interest.
The product is subject to the following: Abcam Restricted Use Statement