Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-methoxycoumarin-3-carboxylic acid | 20300-59-8 | 97.0% | 220.18 g/mol | C11H8O5 | 25 G
SDP

Catalog No. 5000303712
Encompass_Preferred

7-Methoxycoumarin-3-carboxylic acid is a coumarin-based fluorescent dye used for labeling peptides and synthesizing fluorescent probes. It provides ultraviolet excitation and blue emission suitable for fluorescence assays, probe development, and HPLC derivatization. The material is supplied at research-grade purity for laboratory use.

  • High purity (97.0%).
  • Excitation 345 nm, emission 450 nm.
  • Molecular weight 220.18 g/mol.
  • Chemical formula C11H8O5.
  • Available in multiple pack sizes including 25 g.

Catalog No. 50-003-03712 Supplier Medchemexpress LLC Supplier No. 25GHYW039519
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Methoxycoumarin-3-carboxylic acid is a coumarin-based fluorescent dye used for labeling peptides and synthesizing fluorescent probes. It provides ultraviolet excitation and blue emission suitable for fluorescence assays, probe development, and HPLC derivatization. The material is supplied at research-grade purity for laboratory use.

  • High purity (97.0%).
  • Excitation 345 nm, emission 450 nm.
  • Molecular weight 220.18 g/mol.
  • Chemical formula C11H8O5.
  • Available in multiple pack sizes including 25 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.