Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-methoxycoumarin-3-carboxylic acid | 20300-59-8 | MFCD00452770 | ≥97.0% | 220.18 | C11H8O5 | 1 G
SDP

Catalog No. 5000299748
Encompass_Preferred

7-Methoxycoumarin-3-carboxylic acid is a coumarin-derived fluorescent dye used to label peptides and other biomolecules for biochemical assays and HPLC derivatization. It is supplied as a solid with high purity and is prepared as a DMSO stock for use in fluorescence-based assays; handle and store protected from light.

  • Excitation and emission in the near-UV/blue range (Ex ≈345-355 nm, Em ≈405-450 nm).
  • Suitable for peptide labeling and HPLC derivatization.
  • High purity appropriate for analytical applications (~97.0%).
  • Soluble in DMSO (≈8 mg/mL with sonication and warming).
  • Store protected from light; in solution, keep frozen for extended stability.

Catalog No. 50-002-99748 Supplier Medchemexpress LLC Supplier No. HYW0395191G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Methoxycoumarin-3-carboxylic acid is a coumarin-derived fluorescent dye used to label peptides and other biomolecules for biochemical assays and HPLC derivatization. It is supplied as a solid with high purity and is prepared as a DMSO stock for use in fluorescence-based assays; handle and store protected from light.

  • Excitation and emission in the near-UV/blue range (Ex ≈345-355 nm, Em ≈405-450 nm).
  • Suitable for peptide labeling and HPLC derivatization.
  • High purity appropriate for analytical applications (~97.0%).
  • Soluble in DMSO (≈8 mg/mL with sonication and warming).
  • Store protected from light; in solution, keep frozen for extended stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.