Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-methoxycoumarin-3-carboxylic acid | 20300-59-8 | MFCD00452770 | 97.0% | 220.18 | C11H8O5 | 250 MG
SDP

Catalog No. 5000303461
Encompass_Preferred

7-Methoxycoumarin-3-carboxylic acid is a coumarin-based fluorescent dye used to derivatize molecules for fluorescence detection in biochemical assays. It is commonly applied to label peptides, for HPLC derivatization, and for development of fluorescent probes.

  • Fluorescent coumarin dye with characteristic excitation and emission peaks.
  • Excitation ~345-355 nm; emission ~405-450 nm.
  • High purity (manufacturer lists 97.0%).
  • Chemical formula C11H8O5; molecular weight 220.18.
  • Available in multiple sizes from 250 mg to 50 g.
  • Suitable for peptide labeling and HPLC derivatization.
  • Supplied with data sheet and safety data sheet (SDS).

Catalog No. 50-003-03461 Supplier Medchemexpress LLC Supplier No. 250MGHYW039519
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Methoxycoumarin-3-carboxylic acid is a coumarin-based fluorescent dye used to derivatize molecules for fluorescence detection in biochemical assays. It is commonly applied to label peptides, for HPLC derivatization, and for development of fluorescent probes.

  • Fluorescent coumarin dye with characteristic excitation and emission peaks.
  • Excitation ~345-355 nm; emission ~405-450 nm.
  • High purity (manufacturer lists 97.0%).
  • Chemical formula C11H8O5; molecular weight 220.18.
  • Available in multiple sizes from 250 mg to 50 g.
  • Suitable for peptide labeling and HPLC derivatization.
  • Supplied with data sheet and safety data sheet (SDS).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.