Learn More
Medchemexpress LLC 7-Methoxycoumarin-3-carboxylic acid | 20300-59-8 | MFCD00452770 | 97.0% | 220.18 g/mol | C11H8O5 | 10 G
SDP
Catalog No. 5000301917
Encompass_Preferred
7-Methoxycoumarin-3-carboxylic acid is a small-molecule fluorescent dye that absorbs in the near-UV (≈345 nm) and emits blue fluorescence (≈450 nm). Supplied as a white to off-white solid, it is used for labeling peptides, HPLC derivatization, and developing FRET-based probes in biochemical assays. It is soluble in DMSO (~8 mg/mL) and typically supplied at 97% purity.
Catalog No. 50-003-01917 Supplier Medchemexpress LLC Supplier No. HYW03951910G
7-Methoxycoumarin-3-carboxylic acid is a small-molecule fluorescent dye that absorbs in the near-UV (≈345 nm) and emits blue fluorescence (≈450 nm). Supplied as a white to off-white solid, it is used for labeling peptides, HPLC derivatization, and developing FRET-based probes in biochemical assays. It is soluble in DMSO (~8 mg/mL) and typically supplied at 97% purity.