Learn More
Medchemexpress LLC 7-methoxycoumarin-3-carboxylic acid | 20300-59-8 | MFCD00452770 | 97.0% | 220.18 g/mol | C11H8O5 | 500 MG
SDP
Catalog No. 5000301695
Encompass_Preferred
7-Methoxycoumarin-3-carboxylic acid is a coumarin-based fluorescent dye used to label peptides and small biomolecules. It exhibits ultraviolet excitation and blue emission, making it suitable for fluorescence assays, probe synthesis, and HPLC derivatization. The compound is supplied as a solid and should be handled and stored protected from light.
Catalog No. 50-003-01695 Supplier Medchemexpress LLC Supplier No. HYW039519500MG
7-Methoxycoumarin-3-carboxylic acid is a coumarin-based fluorescent dye used to label peptides and small biomolecules. It exhibits ultraviolet excitation and blue emission, making it suitable for fluorescence assays, probe synthesis, and HPLC derivatization. The compound is supplied as a solid and should be handled and stored protected from light.