Learn More
Medchemexpress LLC 7-methoxycoumarin-3-carboxylic acid N-succinimidyl ester | 150321-92-9 | MFCD00467593 | ≥97.0% | 10 MG
SDP
Catalog No. 5000297840
Encompass_Preferred
7-Methoxycoumarin-3-carboxylic acid, succinimidyl ester is a coumarin-derived, amine-reactive fluorescent labeling reagent used to conjugate peptides and proteins for blue/UV fluorescence detection and intracellular imaging applications. The NHS ester enables efficient coupling to primary amines under mild conditions.
  • Amine-reactive N-hydroxysuccinimide (NHS) ester for efficient conjugation.
  • Strong blue fluorescence suitable for labeling peptides and proteins.
  • Useful for synthesis of hydrophilic dye probes and intracellular viscosity measurements.
  • Available in small research pack sizes for mg-scale conjugation experiments.
  • Typical purity ≥97% (HPLC); consult the certificate of analysis for batch-specific data.
Catalog No. 50-002-97840 Supplier Medchemexpress LLC Supplier No. HYD108310MG
7-Methoxycoumarin-3-carboxylic acid, succinimidyl ester is a coumarin-derived, amine-reactive fluorescent labeling reagent used to conjugate peptides and proteins for blue/UV fluorescence detection and intracellular imaging applications. The NHS ester enables efficient coupling to primary amines under mild conditions.
  • Amine-reactive N-hydroxysuccinimide (NHS) ester for efficient conjugation.
  • Strong blue fluorescence suitable for labeling peptides and proteins.
  • Useful for synthesis of hydrophilic dye probes and intracellular viscosity measurements.
  • Available in small research pack sizes for mg-scale conjugation experiments.
  • Typical purity ≥97% (HPLC); consult the certificate of analysis for batch-specific data.