Learn More
Medchemexpress LLC 7-Methoxycoumarin-3-carboxylic acid, SE | 150321-92-9 | 317.25 | 100 MG
SDP
Catalog No. 5000431338
Encompass_Preferred
7-Methoxycoumarin-3-carboxylic acid, SE can be used for the synthesis of hydrophilic dye compound 12. Compound 12 shows a homogeneous distribution inside the cell and represents a suitable probe for viscosity measurements in the cytoplasm. This product is for research purposes only and not for sale to patients.
  • Used for the synthesis of hydrophilic dye compound 12
  • Suitable probe for viscosity measurements in the cytoplasm
Catalog No. 50-004-31338 Supplier Medchemexpress LLC Supplier No. HYD1083100MG
7-Methoxycoumarin-3-carboxylic acid, SE can be used for the synthesis of hydrophilic dye compound 12. Compound 12 shows a homogeneous distribution inside the cell and represents a suitable probe for viscosity measurements in the cytoplasm. This product is for research purposes only and not for sale to patients.
  • Used for the synthesis of hydrophilic dye compound 12
  • Suitable probe for viscosity measurements in the cytoplasm