Learn More
Medchemexpress LLC 7-Methoxycoumarin-3-carboxylic acid, SE | 150321-92-9 | 317.25 | 5 MG
SDP
Catalog No. 5000428554
Encompass_Preferred
7-Methoxycoumarin-3-carboxylic acid, SE can be used for the synthesis of hydrophilic dye compound 12. Compound 12 shows a homogeneous distribution inside the cell and represents a suitable probe for viscosity measurements in the cytoplasm.
Catalog No. 50-004-28554 Supplier Medchemexpress LLC Supplier No. HYD10835MG
Add to Cart
Add to Cart
7-Methoxycoumarin-3-carboxylic acid, SE can be used for the synthesis of hydrophilic dye compound 12. Compound 12 shows a homogeneous distribution inside the cell and represents a suitable probe for viscosity measurements in the cytoplasm.