Learn More
Medchemexpress LLC 7-methoxycoumarin-4-acetic acid | 62935-72-2 | MFCD00009774 | 98.9% | 234.21 g/mol | C12H10O5 | 250 MG
SDP
Catalog No. 5000303652
Encompass_Preferred
MCA (7-methoxycoumarin-4-acetic acid) is a coumarin-derived fluorescent reagent used to label peptides and to develop FRET peptide substrates for measuring protease and enzyme activity. It is supplied as a white to off-white solid with high chemical purity and is suitable for small-scale laboratory research.
  • High purity suitable for sensitive fluorescence assays.
  • Functions as a fluorescent leaving group in FRET substrates.
  • White to off-white solid, easy to handle in laboratory workflows.
  • Available in small laboratory pack sizes for research use.
  • Stable when stored protected from light at recommended temperatures.
Catalog No. 50-003-03652 Supplier Medchemexpress LLC Supplier No. 250MGHYW027544
MCA (7-methoxycoumarin-4-acetic acid) is a coumarin-derived fluorescent reagent used to label peptides and to develop FRET peptide substrates for measuring protease and enzyme activity. It is supplied as a white to off-white solid with high chemical purity and is suitable for small-scale laboratory research.
  • High purity suitable for sensitive fluorescence assays.
  • Functions as a fluorescent leaving group in FRET substrates.
  • White to off-white solid, easy to handle in laboratory workflows.
  • Available in small laboratory pack sizes for research use.
  • Stable when stored protected from light at recommended temperatures.