Learn More
Cayman Chemical 7-Methoxyflavone, Purity: ≥98%, CAS: 22395-22-8, 10 g
SDP
Catalog No. 502950229
Encompass_Preferred
A flavone; inhibits aromatase in a cell-free assay (IC50 = 1.9 µM); inhibits LPS-induced NO production by 17.74% in RAW 264.7 macrophages at 20 µM; activates androgen and/or glucocorticoid receptor transcriptional activity in a reporter assay; has antinociceptive activity in the acetic acid-induced writhing test in mice (ED50 = 82.5 µmol/kg)
Catalog No. 50-295-0229 Supplier Cayman Chemical Supplier No. 3349710 G
A flavone; inhibits aromatase in a cell-free assay (IC50 = 1.9 µM); inhibits LPS-induced NO production by 17.74% in RAW 264.7 macrophages at 20 µM; activates androgen and/or glucocorticoid receptor transcriptional activity in a reporter assay; has antinociceptive activity in the acetic acid-induced writhing test in mice (ED50 = 82.5 µmol/kg)