Learn More
Medchemexpress LLC 7-Methoxyflavone | 22395-22-8 | 1 ML
SDP
Catalog No. 5000418212
Encompass_Preferred
7-Methoxyflavone is a flavonoid compound isolated from Zornia brasiliensis. It binds to human serum albumin and acts as an aromatase inhibitor. This compound demonstrates peripheral analgesic and certain antitumor activities, specifically effective against chemical pain but not thermal pain.
  • Flavonoid compound
  • Isolated from Zornia brasiliensis
  • Binds to human serum albumin
  • Acts as an aromatase inhibitor
  • Exhibits peripheral analgesic activity
  • Shows certain antitumor activities
  • Effective against chemical pain
Catalog No. 50-004-18212 Supplier Medchemexpress LLC Supplier No. HYN848710MM/1ML
7-Methoxyflavone is a flavonoid compound isolated from Zornia brasiliensis. It binds to human serum albumin and acts as an aromatase inhibitor. This compound demonstrates peripheral analgesic and certain antitumor activities, specifically effective against chemical pain but not thermal pain.
  • Flavonoid compound
  • Isolated from Zornia brasiliensis
  • Binds to human serum albumin
  • Acts as an aromatase inhibitor
  • Exhibits peripheral analgesic activity
  • Shows certain antitumor activities
  • Effective against chemical pain