Learn More
Medchemexpress LLC 7-Methoxyflavone | 22395-22-8 | MFCD00017462 | 100 MG
SDP
Catalog No. 5000419664
Encompass_Preferred
7-Methoxyflavone is a flavonoid compound isolated from Zornia brasiliensis. It binds to human serum albumin and acts as an aromatase inhibitor. It possesses peripheral analgesic and certain antitumor activities, being effective against chemical pain but not thermal pain.
  • Isolated from Zornia brasiliensis
  • Binds to human serum albumin
  • Acts as an aromatase inhibitor
  • Exhibits peripheral analgesic activity
  • Exhibits antitumor activities
  • Effective against chemical pain
Catalog No. 50-004-19664 Supplier Medchemexpress LLC Supplier No. HYN8487100MG
7-Methoxyflavone is a flavonoid compound isolated from Zornia brasiliensis. It binds to human serum albumin and acts as an aromatase inhibitor. It possesses peripheral analgesic and certain antitumor activities, being effective against chemical pain but not thermal pain.
  • Isolated from Zornia brasiliensis
  • Binds to human serum albumin
  • Acts as an aromatase inhibitor
  • Exhibits peripheral analgesic activity
  • Exhibits antitumor activities
  • Effective against chemical pain