Learn More
Medchemexpress LLC HY-N6631 100mg Medchemexpress, 7-Methoxyisoflavone CAS:1621-56-3 Purity:>98%
SDP
Catalog No. 501933965
Encompass_Preferred
Medchemexpress, HY-N6631 100mg 7-Methoxyisoflavone CAS:1621-56-3 7-Methoxyisoflavone is an isoflavone derivative and also an activator of adenosine monophosphate-activated protein kinase ( AMPK ). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3965 Supplier Medchemexpress LLC Supplier No. HYN6631100MG
Medchemexpress, HY-N6631 100mg 7-Methoxyisoflavone CAS:1621-56-3 7-Methoxyisoflavone is an isoflavone derivative and also an activator of adenosine monophosphate-activated protein kinase ( AMPK ). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.