Learn More
Medchemexpress LLC 7-Methoxyisoflavone | 1621-56-3 | 250 MG
SDP
Catalog No. 5000422016
Encompass_Preferred
7-Methoxyisoflavone is an isoflavone derivative that acts as an activator of adenosine monophosphate-activated protein kinase (AMPK). It is suitable for research applications, preventing the serum-induced decrease in AMPK phosphorylation in serum-starved cells treated with 10% FBS.
  • Activates adenosine monophosphate-activated protein kinase (AMPK)
  • Prevents serum-induced decrease in AMPK phosphorylation
  • High purity of 99.47%
  • White to off-white solid appearance
  • Soluble in DMSO
  • Available with in vivo dissolution protocols
Catalog No. 50-004-22016 Supplier Medchemexpress LLC Supplier No. 250MGHYN6631
7-Methoxyisoflavone is an isoflavone derivative that acts as an activator of adenosine monophosphate-activated protein kinase (AMPK). It is suitable for research applications, preventing the serum-induced decrease in AMPK phosphorylation in serum-starved cells treated with 10% FBS.
  • Activates adenosine monophosphate-activated protein kinase (AMPK)
  • Prevents serum-induced decrease in AMPK phosphorylation
  • High purity of 99.47%
  • White to off-white solid appearance
  • Soluble in DMSO
  • Available with in vivo dissolution protocols