Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene

Catalog No. 3595055ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-950-55ML 5 mL
35-950-51ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 35-950-55ML Supplier Sigma Aldrich Supplier No. 3595055ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 7-methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene is a soluble amine base
  • It forms 1:1 complexes with 4-nitrophenyl[bis(diethylsulfonyl)]methane and phenyl[bis(diethylsulfonyl)]methane and their structures have been studied by ft-IR, 1h NMR and pm5 semiempirical methods
  • Catalytic performance of 7-methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene for the transesterification of rapseed oil with methanol has been investigated.7-methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene may be used in microwave-promoted, palladium-catalyzed c-n bond-forming reactions with aryl/heteroaryl nonaflates and amines
  • It may be used as strong base to investigate the podand solvents, tris(oxaalkyl)phenylsilanes and tris(oxaalkyl)phosphates

Specifications

CAS 84030-20-6
Density 1.067 g/mL (at 25°C (literature))
Boiling Point 75°C to 79°C (0.1 mmHg)
Molecular Formula C8H15N3
Refractive Index n20/D 1.537 (literature)
Linear Formula C8H15N3
MDL Number MFCD00043004
Quantity 5 mL
Synonym 1,3,4,6,7,8-Hexahydro-1-methyl-2 H-pyrimido[1,2-a]pyrimidine; MTBD
Molecular Weight (g/mol) 153.22
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.