Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene | 84030-20-6 | MFCD00043004 | 99.2% | C8H15N3 | 500 MG
SDP

Catalog No. 5000434730
Encompass_Preferred

7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene is a drug intermediate used for the synthesis of various active compounds. This product is intended for research use only.

  • Purity: 99.2%
  • Molecular weight: 153.23
  • Chemical formula: C8H15N3
  • CAS number: 84030-20-6
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Storage conditions:
    • Pure form: -20°C for 3 years, 4°C for 2 years
    • In solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-34730 Supplier Medchemexpress LLC Supplier No. HY59157500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene is a drug intermediate used for the synthesis of various active compounds. This product is intended for research use only.

  • Purity: 99.2%
  • Molecular weight: 153.23
  • Chemical formula: C8H15N3
  • CAS number: 84030-20-6
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Storage conditions:
    • Pure form: -20°C for 3 years, 4°C for 2 years
    • In solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.