Learn More
Medchemexpress LLC 7-methyl-1,5-benzodioxepin-3-one (watermelon ketone) | 28940-11-6 | MFCD07371373 | 100.0% | 178.18 g/mol | C10H10O3 | 1 ML
SDP
Catalog No. 5000303202
Encompass_Preferred
Watermelon ketone (7-methyl-1,5-benzodioxepin-3-one; CAS 28940-11-6) is a fragrance and flavor compound with a fresh marine/watermelon aroma. It is used in perfumery, flavor research, and formulation studies, and is provided as a high-purity reagent and small-volume pre-diluted solutions for laboratory use.
  • High purity for analytical and formulation work (listed as 99.95%).
  • Molecular formula C10H10O3 and molecular weight 178.18 g/mol.
  • Available as neat material or small-volume pre-diluted solutions for convenience.
  • Commonly used in perfumery, fragrance formulation, and flavor research.
  • Supplied in liquid form with solubility in common organic solvents.
Catalog No. 50-003-03202 Supplier Medchemexpress LLC Supplier No. HYW01662210MM/1ML
Watermelon ketone (7-methyl-1,5-benzodioxepin-3-one; CAS 28940-11-6) is a fragrance and flavor compound with a fresh marine/watermelon aroma. It is used in perfumery, flavor research, and formulation studies, and is provided as a high-purity reagent and small-volume pre-diluted solutions for laboratory use.
  • High purity for analytical and formulation work (listed as 99.95%).
  • Molecular formula C10H10O3 and molecular weight 178.18 g/mol.
  • Available as neat material or small-volume pre-diluted solutions for convenience.
  • Commonly used in perfumery, fragrance formulation, and flavor research.
  • Supplied in liquid form with solubility in common organic solvents.