Learn More
Medchemexpress LLC 7-Methyl-1-naphthyl acetic acid | 6836-22-2 | 216.24 | 1 G
SDP
Catalog No. 5000445751
Encompass_Preferred
7-Methyl-1-naphthyl acetic acid is a potent inhibitor of auxin action in plants. It inhibits AUX1, PIN, and ABCB protein-mediated auxin transport expressed in yeast. This compound can be used in auxin-dependent development research.
  • Potent inhibitor of auxin action in plants
  • Inhibits AUX1, PIN, and ABCB protein-mediated auxin transport
  • Suitable for auxin-dependent development research
Catalog No. 50-004-45751 Supplier Medchemexpress LLC Supplier No. HYW0038361G
7-Methyl-1-naphthyl acetic acid is a potent inhibitor of auxin action in plants. It inhibits AUX1, PIN, and ABCB protein-mediated auxin transport expressed in yeast. This compound can be used in auxin-dependent development research.
  • Potent inhibitor of auxin action in plants
  • Inhibits AUX1, PIN, and ABCB protein-mediated auxin transport
  • Suitable for auxin-dependent development research