Learn More
Medchemexpress LLC 7-methyl-5-[3-(piperazin-1-ylmethyl)-1,2,4-oxadiazol-5-yl]-2-[[4-(trifluoromethoxy)phenyl]methyl]-3H-isoindol-1-... | 1092453-15-0 | MFCD31657360 | 98.0% | 487.47 g/mol | C24H24F3N5O3 | 25 MG
SDP
Catalog No. 5000239748
Encompass_Preferred
AZD-8529 is a research-stage small-molecule positive allosteric modulator (PAM) of the metabotropic glutamate receptor 2 (mGluR2). It is used in preclinical pharmacology and behavioral studies to probe mGluR2-mediated signaling and pharmacodynamics. Supplied as a high-purity solid for laboratory research.
Catalog No. 50-002-39748 Supplier Medchemexpress LLC Supplier No. HY10745725MG
AZD-8529 is a research-stage small-molecule positive allosteric modulator (PAM) of the metabotropic glutamate receptor 2 (mGluR2). It is used in preclinical pharmacology and behavioral studies to probe mGluR2-mediated signaling and pharmacodynamics. Supplied as a high-purity solid for laboratory research.